4-[(2-Bromoacetyl)amino]-N,N-dipropylbenzamide

Supplier: Matrix Scientific

035043-500MG
101838-874EA 306.15 USD
101838-874
4-[(2-Bromoacetyl)amino]-N,N-dipropylbenzamide
4-[(2-Bromoacetyl)amino]-N,N-dipropylbenzamide

Hazard: Irritant

Formula: C₁₅H₂₁BrN₂O₂
MDL Number: MFCD12026621
CAS Number: 1138442-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR