3-[(2-Bromoacetyl)amino]-N,N-diethylbenzamide

Supplier: Matrix Scientific

035040-500MG
101838-868EA 306.15 USD
101838-868
3-[(2-Bromoacetyl)amino]-N,N-diethylbenzamide
3-[(2-Bromoacetyl)amino]-N,N-diethylbenzamide

Hazard: Irritant

Formula: C₁₃H₁₇BrN₂O₂
MDL Number: MFCD12026618
CAS Number: 1138442-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR