2-Bromo-N-[3-(2-methoxyethoxy)phenyl]acetamide

Supplier: Matrix Scientific

035026-500MG
101838-840EA 306.15 USD
101838-840
2-Bromo-N-[3-(2-methoxyethoxy)phenyl]acetamide
2-Bromo-N-[3-(2-methoxyethoxy)phenyl]acetamide

Hazard: Irritant

Formula: C₁₁H₁₄BrNO₃
MDL Number: MFCD12026606
CAS Number: 1138442-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR