2-Bromo-N-[3-(sec-butoxy)phenyl]acetamide

Supplier: Matrix Scientific

035025-500MG
101838-838EA 306.15 USD
101838-838
2-Bromo-N-[3-(sec-butoxy)phenyl]acetamide
2-Bromo-N-[3-(sec-butoxy)phenyl]acetamide

Hazard: Irritant

Formula: C₁₂H₁₆BrNO₂
MDL Number: MFCD12026605
CAS Number: 1138442-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR