2-Bromo-N-(3-isopropoxyphenyl)acetamide

Supplier: Matrix Scientific

035023-500MG
101838-834EA 306.15 USD
101838-834
2-Bromo-N-(3-isopropoxyphenyl)acetamide
2-Bromo-N-(3-isopropoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₁H₁₄BrNO₂
MDL Number: MFCD12026603
CAS Number: 918408-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR