2-Bromo-N-butylacetamide

035021-500MG
101838-830EA 215.47 USD
101838-830
2-Bromo-N-butylacetamide
2-Bromo-N-butylacetamide

Hazard: Irritant

Formula: C₆H₁₂BrNO
MDL Number: MFCD12026601
CAS Number: 67056-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR