2-Bromo-N-[2-(3-phenylpropoxy)phenyl]acetamide

Supplier: Matrix Scientific

035011-500MG
101838-810EA 306.15 USD
101838-810
2-Bromo-N-[2-(3-phenylpropoxy)phenyl]acetamide
2-Bromo-N-[2-(3-phenylpropoxy)phenyl]acetamide

Hazard: Irritant

Formula: C₁₇H₁₈BrNO₂
MDL Number: MFCD12026591
CAS Number: 1138442-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR