2-Bromo-N-(3-ethoxyphenyl)acetamide

Supplier: Matrix Scientific

035003-500MG
101838-794EA 306.15 USD
101838-794
2-Bromo-N-(3-ethoxyphenyl)acetamide
2-Bromo-N-(3-ethoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₂BrNO₂
MDL Number: MFCD12026583
CAS Number: 34325-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR