N-[2-(Allyloxy)phenyl]-2-bromoacetamide

Supplier: Matrix Scientific

035002-500MG
101838-792EA 306.15 USD
101838-792
N-[2-(Allyloxy)phenyl]-2-bromoacetamide
N-[2-(Allyloxy)phenyl]-2-bromoacetamide

Hazard: Irritant

Formula: C₁₁H₁₂BrNO₂
MDL Number: MFCD12026582
CAS Number: 1138442-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR