2-Bromo-N-[3-(isopentyloxy)phenyl]acetamide

Supplier: Matrix Scientific

035000-500MG
101838-788EA 306.15 USD
101838-788
2-Bromo-N-[3-(isopentyloxy)phenyl]acetamide
2-Bromo-N-[3-(isopentyloxy)phenyl]acetamide

Hazard: Irritant

Formula: C₁₃H₁₈BrNO₂
MDL Number: MFCD12026580
CAS Number: 1138442-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR