sec-Butyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate

Supplier: Matrix Scientific

034999-500MG
101838-786EA 306.15 USD
101838-786
sec-Butyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate
sec-Butyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate

Hazard: Irritant

Formula: C₁₂H₁₉BrN₂O₄
MW: 335.21 g/mol
MDL Number: MFCD12026579
CAS Number: 1219147-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR