2-Bromo-N-[3-(hexyloxy)phenyl]acetamide

Supplier: Matrix Scientific

034996-500MG
101838-780EA 306.15 USD
101838-780
2-Bromo-N-[3-(hexyloxy)phenyl]acetamide
2-Bromo-N-[3-(hexyloxy)phenyl]acetamide

Hazard: Irritant

Formula: C₁₄H₂₀BrNO₂
MDL Number: MFCD12026576
CAS Number: 1138442-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR