Decyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate

Supplier: Matrix Scientific

034995-500MG
101838-778EA 306.15 USD
101838-778
Decyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate
Decyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate

Hazard: Irritant

Formula: C₁₈H₃₁BrN₂O₄
MDL Number: MFCD12026575
CAS Number: 1219219-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR