2-Bromo-N-(2-isopropylphenyl)acetamide

Supplier: Matrix Scientific

034994-500MG
101838-776EA 306.15 USD
101838-776
2-Bromo-N-(2-isopropylphenyl)acetamide
2-Bromo-N-(2-isopropylphenyl)acetamide

Hazard: Irritant

Formula: C₁₁H₁₄BrNO
MDL Number: MFCD02974376
CAS Number: 349120-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR