Dodecyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate

Supplier: Matrix Scientific

034990-500MG
101838-768EA 306.15 USD
101838-768
Dodecyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate
Dodecyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate

Hazard: Irritant

Formula: C₂₀H₃₅BrN₂O₄
MDL Number: MFCD12026571
CAS Number: 1219298-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR