Isopentyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate

Supplier: Matrix Scientific

034988-500MG
101838-764EA 306.15 USD
101838-764
Isopentyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate
Isopentyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate

Hazard: Irritant

Formula: C₁₃H₂₁BrN₂O₄
MDL Number: MFCD12026569
CAS Number: 1219426-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR