2-Bromo-N-(3-isobutoxyphenyl)acetamide

Supplier: Matrix Scientific

034987-500MG
101838-762EA 306.15 USD
101838-762
2-Bromo-N-(3-isobutoxyphenyl)acetamide
2-Bromo-N-(3-isobutoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₆BrNO₂
MDL Number: MFCD12026568
CAS Number: 1138442-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR