2-Bromo-N-[4-(2-methoxyethoxy)phenyl]acetamide

Supplier: Matrix Scientific

034982-500MG
101838-752EA 306.15 USD
101838-752
2-Bromo-N-[4-(2-methoxyethoxy)phenyl]acetamide
2-Bromo-N-[4-(2-methoxyethoxy)phenyl]acetamide

Hazard: Irritant

Formula: C₁₁H₁₄BrNO₃
MDL Number: MFCD12026563
CAS Number: 1138445-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR