2-Bromo-N-[4-(heptyloxy)phenyl]acetamide

Supplier: Matrix Scientific

034975-500MG
101838-738EA 306.15 USD
101838-738
2-Bromo-N-[4-(heptyloxy)phenyl]acetamide
2-Bromo-N-[4-(heptyloxy)phenyl]acetamide

Hazard: Irritant

Formula: C₁₅H₂₂BrNO₂
MDL Number: MFCD12026556
CAS Number: 1138445-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR