2-Bromo-N-(4-butoxyphenyl)acetamide

Supplier: Matrix Scientific

034971-500MG
101838-730EA 306.15 USD
101838-730
2-Bromo-N-(4-butoxyphenyl)acetamide
2-Bromo-N-(4-butoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₆BrNO₂
MDL Number: MFCD12026553
CAS Number: 195393-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR