2-Bromo-N-[4-(hexyloxy)phenyl]acetamide

Supplier: Matrix Scientific

034970-500MG
101838-728EA 306.15 USD
101838-728
2-Bromo-N-[4-(hexyloxy)phenyl]acetamide
2-Bromo-N-[4-(hexyloxy)phenyl]acetamide

Hazard: Irritant

Formula: C₁₄H₂₀BrNO₂
MDL Number: MFCD12026552
CAS Number: 1138445-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR