2-[(2-Bromoacetyl)amino]-N,N-dimethylbenzamide

Supplier: Matrix Scientific

034968-500MG
101838-724EA 306.15 USD
101838-724
2-[(2-Bromoacetyl)amino]-N,N-dimethylbenzamide
2-[(2-Bromoacetyl)amino]-N,N-dimethylbenzamide

Hazard: Irritant

Formula: C₁₁H₁₃BrN₂O₂
MDL Number: MFCD12026550
CAS Number: 1138445-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR