N-Benzyl-2-[(2-bromoacetyl)amino]-N-methylbenzamide

Supplier: Matrix Scientific

034964-500MG
101838-716EA 306.15 USD
101838-716
N-Benzyl-2-[(2-bromoacetyl)amino]-N-methylbenzamide
N-Benzyl-2-[(2-bromoacetyl)amino]-N-methylbenzamide

Hazard: Irritant

Formula: C₁₇H₁₇BrN₂O₂
MDL Number: MFCD12026546
CAS Number: 1138445-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR