2-Phenoxyethyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate
Supplier: Matrix Scientific
034961-500MG
101838-710EA
306.15
USD
101838-710
2-Phenoxyethyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate
2-Phenoxyethyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate
Hazard: Irritant
Formula:
C₁₆H₁₉BrN₂O₅ |
MDL Number:
MFCD12026543 CAS Number: 1219272-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...