2-Bromo-N-[2-(1-pyrrolidinylcarbonyl)phenyl]acetamide

Supplier: Matrix Scientific

034959-500MG
101838-706EA 306.15 USD
101838-706
2-Bromo-N-[2-(1-pyrrolidinylcarbonyl)phenyl]acetamide
2-Bromo-N-[2-(1-pyrrolidinylcarbonyl)phenyl]acetamide

Hazard: Irritant

Formula: C₁₃H₁₅BrN₂O₂
MW: 311.19 g/mol
MDL Number: MFCD12026541
CAS Number: 1138445-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR