Propyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate

Supplier: Matrix Scientific

034958-500MG
101838-704EA 306.15 USD
101838-704
Propyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate
Propyl-2-[1-(2-bromoacetyl)-3-oxo-2-piperazinyl]acetate

Hazard: Irritant

Formula: C₁₁H₁₇BrN₂O₄
MW: 321.18 g/mol
MDL Number: MFCD12026540
CAS Number: 1219402-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR