2-Bromo-N-[4-(2,3-dihydro-1H-indol-1-ylsulfonyl)phenyl]acetamide

Supplier: Matrix Scientific

034944-500MG
101838-676EA 306.15 USD
101838-676
2-Bromo-N-[4-(2,3-dihydro-1H-indol-1-ylsulfonyl)phenyl]acetamide
2-Bromo-N-[4-(2,3-dihydro-1H-indol-1-ylsulfonyl)phenyl]acetamide

Hazard: Irritant

Formula: C₁₆H₁₅BrN₂O₃S
MDL Number: MFCD12026529
CAS Number: 1138445-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR