2-Bromo-N-heptylacetamide

Supplier: Matrix Scientific

034943-500MG
101838-674EA 306.15 USD
101838-674
2-Bromo-N-heptylacetamide
2-Bromo-N-heptylacetamide

Hazard: Irritant

Formula: C₉H₁₈BrNO
MDL Number: MFCD12026528
CAS Number: 5463-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR