2-Bromo-N-(2-methyl-4-nitrophenyl)acetamide

Supplier: Matrix Scientific

034940-500MG
101838-668EA 306.15 USD
101838-668
2-Bromo-N-(2-methyl-4-nitrophenyl)acetamide
2-Bromo-N-(2-methyl-4-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₉H₉BrN₂O₃
MDL Number: MFCD02974370
CAS Number: 57339-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR