2-Bromo-N-(2-methyl-3-nitrophenyl)acetamide

Supplier: Matrix Scientific

034939-500MG
101838-666EA 306.15 USD
101838-666
2-Bromo-N-(2-methyl-3-nitrophenyl)acetamide
2-Bromo-N-(2-methyl-3-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₉H₉BrN₂O₃
MDL Number: MFCD12026526
CAS Number: 1138445-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR