2-Bromo-N-(2-methoxy-5-nitrophenyl)acetamide

Supplier: Matrix Scientific

034938-500MG
101838-664EA 306.15 USD
101838-664
2-Bromo-N-(2-methoxy-5-nitrophenyl)acetamide
2-Bromo-N-(2-methoxy-5-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₉H₉BrN₂O₄
MDL Number: MFCD02974399
CAS Number: 117215-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR