2-Bromo-N-(2-methyl-5-nitrophenyl)acetamide

Supplier: Matrix Scientific

034936-500MG
101838-660EA 306.15 USD
101838-660
2-Bromo-N-(2-methyl-5-nitrophenyl)acetamide
2-Bromo-N-(2-methyl-5-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₉H₉BrN₂O₃
MDL Number: MFCD02974401
CAS Number: 349121-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR