2-Bromo-N-(4-iodophenyl)acetamide

Supplier: Matrix Scientific

034933-500MG
101838-654EA 306.15 USD
101838-654
2-Bromo-N-(4-iodophenyl)acetamide
2-Bromo-N-(4-iodophenyl)acetamide

Hazard: Irritant

Formula: C₈H₇BrINO
MDL Number: MFCD02974369
CAS Number: 6411-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR