2-Bromo-N-(4-phenoxyphenyl)acetamide

Supplier: Matrix Scientific

034930-500MG
101838-648EA 306.15 USD
101838-648
2-Bromo-N-(4-phenoxyphenyl)acetamide
2-Bromo-N-(4-phenoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₄H₁₂BrNO₂
MDL Number: MFCD02660712
CAS Number: 36160-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR