4-[3-(1-Azepanylcarbonyl)anilino]-4-oxobutanoic acid

Supplier: Matrix Scientific

034919-500MG
101838-626EA 306.15 USD
101838-626
4-[3-(1-Azepanylcarbonyl)anilino]-4-oxobutanoic acid
4-[3-(1-Azepanylcarbonyl)anilino]-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₄
MDL Number: MFCD11089870
CAS Number: 1003688-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR