3-(Methylsulfonyl)-1-propanol

Supplier: Matrix Scientific

Synonyms: 3-(Methylsulfonyl)propan-1-ol3-(Methylsulphonyl)propan-1-ol

034912-500MG
101838-612EA 24.62 USD
101838-612
3-(Methylsulfonyl)-1-propanol
3-(Methylsulfonyl)-1-propanol

Hazard: Irritant

Formula: C₄H₁₀O₃S
MW: 138.19 g/mol
MDL Number: MFCD01696727
CAS Number: 2058-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR