3-Isopropoxypropanenitrile

Supplier: Matrix Scientific

034908-500MG
101838-604EA 306.15 USD
101838-604
3-Isopropoxypropanenitrile
3-Isopropoxypropanenitrile

Hazard: Irritant

Formula: C₆H₁₁NO
Boiling Pt: 82 °C
Density: 0.8969
MDL Number: MFCD00019866
CAS Number: 110-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR