1-(4-Methoxyphenyl)-N-methyl-2-propanamine

Supplier: Matrix Scientific

034907-500MG
101838-602EA 338.46 USD
101838-602
1-(4-Methoxyphenyl)-N-methyl-2-propanamine
1-(4-Methoxyphenyl)-N-methyl-2-propanamine

Hazard: Irritant

Formula: C₁₁H₁₇NO
MDL Number: MFCD01708217
CAS Number: 22331-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR