N-Cyclopentyl-N-propylamine

Supplier: Matrix Scientific

034906-500MG
101838-600EA 306.15 USD
101838-600
N-Cyclopentyl-N-propylamine
N-Cyclopentyl-N-propylamine

Hazard: Irritant

Formula: C₈H₁₇N
MDL Number: MFCD08691712
CAS Number: 39190-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR