N-(4-Bromophenyl)-2,2-dimethylpropanamide

Supplier: Matrix Scientific

Synonyms: 4-Bromopivalamide

034903-500MG
101838-594EA 192.31 USD
101838-594
N-(4-Bromophenyl)-2,2-dimethylpropanamide
N-(4-Bromophenyl)-2,2-dimethylpropanamide

Hazard: Irritant

Formula: C₁₁H₁₄BrNO
MW: 256.15 g/mol
MDL Number: MFCD00124399
CAS Number: 24109-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR