1H-Indole-3-carbaldehyde oxime
Supplier: Matrix Scientific
Synonyms:
1H-Indole-3-carbaldehyde Oxime
, 1H-Indole-3-carboxaldehyde Oxime
034889-500MG
101838-568EA
90.77
USD
101838-568
1H-Indole-3-carbaldehyde oxime
1H-Indole-3-carbaldehyde oxime
Hazard: Irritant
Formula:
C₉H₈N₂O MW: 160.18 g/mol Melting Pt: 197…200 °C |
MDL Number:
MFCD00022737 CAS Number: 2592-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...