Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Cyclohexyl-N-propylamine

Supplier: Matrix Scientific


034888-500MG
101838-566EA 260.23 USD InStock
101838-566
N-Cyclohexyl-N-propylamine
N-Cyclohexyl-N-propylamine
CAS: 3592-81-2
MDL Number: MFCD03210942
Molecular Formula: C9H19N
Molecular Weight: 141,26
Boiling Point: 185°C
Density: 0,84
Flash point: 59.5°C
Hazard: Irritant


Formula: C₉H₁₉N
MW: 141.26 g/mol
Boiling Pt: 185 °C
Density: 0,84
Flash Pt: 59,5 °C
MDL Number: MFCD03210942
CAS Number: 3592-81-2
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty