(4-Isopropoxyphenyl)methanamine hydrochloride

Supplier: Matrix Scientific

034880-500MG
101838-550EA 306.15 USD
101838-550
(4-Isopropoxyphenyl)methanamine hydrochloride
(4-Isopropoxyphenyl)methanamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆ClNO
MDL Number: MFCD11226478
CAS Number: 100131-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR