(5-Chloro-2-isopropoxyphenyl)methanamine hydrochloride

Supplier: Matrix Scientific

034879-500MG
101838-548EA 306.15 USD
101838-548
(5-Chloro-2-isopropoxyphenyl)methanamine hydrochloride
(5-Chloro-2-isopropoxyphenyl)methanamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅Cl₂NO
MDL Number: MFCD11226477
CAS Number: 1135292-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR