1-(3-Pyridinylmethyl)-4-piperidinecarboxylic acid

Supplier: Matrix Scientific

034874-500MG
101838-538EA 306.15 USD
101838-538
1-(3-Pyridinylmethyl)-4-piperidinecarboxylic acid
1-(3-Pyridinylmethyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD03371464
CAS Number: 887442-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR