2-Methylcyclopentanamine hydrochloride

Supplier: Matrix Scientific

034870-500MG
101838-530EA 384.62 USD
101838-530
2-Methylcyclopentanamine hydrochloride
2-Methylcyclopentanamine hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClN
MDL Number: MFCD10703569
CAS Number: 5454-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR