4-Methyl-2-(2-pyridinyl)-5-thiazolecarboxylic acid ≥97%

Supplier: Matrix Scientific

034866-1G
101838-522EA 47.69 USD
101838-522
4-Methyl-2-(2-pyridinyl)-5-thiazolecarboxylic acid ≥97%
4-Methyl-2-(2-pyridinyl)-5-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂S
MDL Number: MFCD01316364
CAS Number: 34418-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR