Bis(4-fluorophenyl)difluoromethane ≥95%

Supplier: Matrix Scientific

034838-1G
101838-470EA 144.62 USD
101838-470
Bis(4-fluorophenyl)difluoromethane ≥95%
Bis(4-fluorophenyl)difluoromethane

Hazard: Irritant

Formula: C₁₃H₈F₄
MDL Number: MFCD11845849
CAS Number: 339-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR