1,1,5-Trifluoroindan ≥95%

Supplier: Matrix Scientific

034837-1G
101838-468EA 150.77 USD
101838-468
1,1,5-Trifluoroindan ≥95%
1,1,5-Trifluoroindan

Hazard: Irritant

Formula: C₉H₇F₃
MDL Number: MFCD11845848
CAS Number: 57584-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR