1-Methyl-4-(pentafluoroethyl)benzene ≥95%

Supplier: Matrix Scientific

034834-1G
101838-462EA 106.15 USD
101838-462
1-Methyl-4-(pentafluoroethyl)benzene ≥95%
1-Methyl-4-(pentafluoroethyl)benzene

Hazard: Irritant

Formula: C₉H₇F₅
MDL Number: MFCD09909908
CAS Number: 117081-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR